accessibility menu, dialog, popup

UserName

CAS RN 348575-88-2

CH3NNH3CONHSS(E/Z)

CAS RN 348575-88-2

IUPAC Name: 5-{[2,5-dimethyl-1-(pyridin-3-yl)-1H-pyrrol-3-yl]methylidene}-2-sulfanylidene-1,3-thiazolidin-4-one
Synonyms: 5-{[2,5-dimethyl-1-(pyridin-3-yl)pyrrol-3-yl]methylidene}-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Weight (g/mol): 315.41
Molecular Formula: C15H13N3OS2
InChi Key: YISGMOZSGOGYOU-UHFFFAOYSA-N
SMILES: CC1=CC(C=C2SC(=S)NC2=O)=C(C)N1C1=CN=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)