accessibility menu, dialog, popup

UserName

NaOHOSOOO

CAS RN 34487-61-1

IUPAC Name: sodium 4-[3-(4-hydroxyphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]benzen-1-olate
Synonyms: sodium 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1λ⁶-benzoxathiol-3-yl]benzenolate
Molecular Weight (g/mol): 376.36
Molecular Formula: C19H13NaO5S
InChi Key: HKHYOKBQJILTEI-UHFFFAOYNA-M
SMILES: [Na+].OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C([O-])C=C1
Molecular Weight (g/mol) 
  • (16)
  • (1)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)
  • (4)
  • (7)
  • (3)
  • (1)
Grade 
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
Color 
  • (3)
  • (1)
  • (1)
  • (3)
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (6)
  • (7)
  • (2)
  • (2)
  • (2)
  • (16)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (7)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)