missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red Sodium Salt, Supelco™

Product Code. 15877870
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15877870 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15877870 Supplier Supelco Supplier No. 1117480005
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Indicator ACS

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 285°C
CAS 34487-61-1
MDL Number MFCD00066901
Synonym Phenol red sodium salt, Phenol red sodium salt
Quantity 5 g
Molecular Weight (g/mol) 376.36
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.