CAS RN 33872-80-9
CAS RN 33872-80-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
150.89
Molecular Formula:
C7H7ClMg
InChi Key:
LQVSLLSEXLZRRH-UHFFFAOYSA-M
SMILES:
[Mg++].[Cl-].Cc1cccc[cH-]1
1
–
1
of
1
results
1
o-Tolylmagnesium chloride, 1.4M (22 wt.%) solution in THF/toluene, AcroSeal™, Thermo Scientific Chemicals
CAS: 33872-80-9 Molecular Formula: C7H7ClMg Molecular Weight (g/mol): 150.89 MDL Number: MFCD00010355 InChI Key: LQVSLLSEXLZRRH-UHFFFAOYSA-M Synonym: o-tolylmagnesium chloride,magnesium, chloro 2-methylphenyl,o-tolylmagnesium chloride solution,2-methylphenylmagnesium chloride,unii-gul3abu3p5,o-tolyl magnesium chloride,gul3abu3p5,chloro-o-tolylmagnesium,o-tolylmagnesiumchloride,2-tolyl magnesium chloride PubChem CID: 118563 IUPAC Name: magnesium;methylbenzene;chloride SMILES: [Mg++].[Cl-].Cc1cccc[cH-]1