accessibility menu, dialog, popup

UserName

CAS RN 3386-97-8

CH2NCS

CAS RN 3386-97-8

IUPAC Name: 4-isothiocyanatobut-1-ene
Synonyms: 1-butene, 4-isothiocyanato-
Molecular Weight (g/mol): 113.18
Molecular Formula: C5H7NS
InChi Key: SKIHGKNFJKJXPX-UHFFFAOYSA-N
SMILES: C=CCCN=C=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results