accessibility menu, dialog, popup

UserName

CAS RN 334-50-9

HHHClClClH2NHNNH2

CAS RN 334-50-9

IUPAC Name: trihydrogen (4-aminobutyl)(3-aminopropyl)amine trichloride
Synonyms: trihydrogen spermidine trichloride
Molecular Weight (g/mol): 254.62
Molecular Formula: C7H22Cl3N3
InChi Key: LCNBIHVSOPXFMR-UHFFFAOYSA-N
SMILES: [H+].[H+].[H+].[Cl-].[Cl-].[Cl-].NCCCCNCCCN
Molecular Weight (g/mol) 
  • (10)
  • (4)
Quantity 
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (1)
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (10)
  • (4)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)