missing translation for 'onlineSavingsMsg'
Learn More

Spermidine Trihydrochloride 99% Crystalline MP Biomedicals

Product Code. 11453150 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11453150

Brand: MP Biomedicals 0210047201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 334-50-9
Assay Percent Range 99%
Molecular Formula C7H22Cl3N3
MDL Number MFCD00012918
Quantity 1 g
Synonym spermidine trihydrochloride, n-3-aminopropyl-1,4-butanediamine trihydrochloride, spermidine hydrochloride, n1-3-aminopropyl butane-1,4-diamine trihydrochloride, unii-1o14bed398, 1,4-butanediamine, n-3-aminopropyl-, trihydrochloride, spermidine, trihydrochloride, 1,4-butanediamine, n-3-aminopropyl , hydrochloride, n-3-aminopropyl butane-1,4-diamine trihydrochloride, 4-azoniaoctamethylenediammonium trichloride
InChI Key LCNBIHVSOPXFMR-UHFFFAOYSA-N
SMILES [H+].[H+].[H+].[Cl-].[Cl-].[Cl-].NCCCCNCCCN
IUPAC Name trihydrogen (4-aminobutyl)(3-aminopropyl)amine trichloride
Molecular Weight (g/mol) 254.62
PubChem CID 9539
Formula Weight 254.6
Physical Form Crystalline
Chemical Name or Material Spermidine Trihydrochloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.