accessibility menu, dialog, popup

UserName

CAS RN 3326-32-7

IUPAC Name: 3',6'-dihydroxy-5-isothiocyanato-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: FITC F5itc fluorescein isothiocyanate isomer 1 fitc isomer 1 fluorescein-5-isothiocyanat fluoreszein-5-isothiocyanat 5-fitc fluorescein 5 isothiocyanate
Molecular Weight (g/mol): 389.38
Molecular Formula: C21H11NO5S
InChi Key: MHMNJMPURVTYEJ-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(OC3=CC(O)=CC=C3C22OC(=O)C3=CC(=CC=C23)N=C=S)=C1
Molecular Weight (g/mol) 
  • (9)
  • (6)
Quantity 
  • (5)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (6)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (6)
  • (2)
  • (5)
  • (9)
  • (6)
  • (5)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (6)
  • (3)
  • (3)