CAS RN 33103-93-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Demethyl Memantine Hydrochloride, TRC
CAS: 33103-93-4 Molecular Formula: C11 H19 N . Cl H Molecular Weight (g/mol): 201.74 Synonym: Tricyclo[3.3.1.13,7]decan-1-amine, 3-methyl-, hydrochloride (1:1),1-Adamantanamine, 3-methyl-, hydrochloride (7CI,8CI),Tricyclo[3.3.1.13,7]decan-1-amine, 3-methyl-, hydrochloride,Demethyl Memantine Hydrochloride,Desmethylmemantine Hydrochloride IUPAC Name: 3-methyladamantan-1-amine;hydrochloride SMILES: Cl.CC12CC3CC(C1)CC(N)(C3)C2
2
3-Methyladamantan-1-amine Hydrochloride (Desmethylmemantine Hydrochloride), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.