missing translation for 'onlineSavingsMsg'
Learn More

3-Methyladamantan-1-amine Hydrochloride (Desmethylmemantine Hydrochloride), Mikromol™

Product Code. 30459585
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459585 4 x 25 mg 100mg
30052026 25 mg 25mg
30051353 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30459585 Supplier Mikromol Supplier No. MM1045.0800254AMP
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-Methyladamantan-1-amine Hydrochloride (Desmethylmemantine Hydrochloride), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Demethyl Memantine Hydrochloride
CAS 33103-93-4
Analyte or Component Names Demethyl Memantine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C11 H19 N . Cl H
Certifications/Compliance ISO 17025
Synonym 3-Methyladamantan-1-amine Hydrochloride, Tricyclo[3.3.1.13,7]decan-1-amine, 3-methyl-, hydrochloride (1:1), 1-Adamantanamine, 3-methyl-, hydrochloride (7CI,8CI), Tricyclo[3.3.1.13,7]decan-1-amine, 3-methyl-, hydrochloride, Demethyl Memantine Hydrochloride, Desmethylmemantine Hydrochloride, Memantine Related Compound G as Hydrochloride
InChI Formula InChI=1S/C11H19N.ClH/c1-10-3-8-2-9(4-10)6-11(12,5-8)7-10;/h8-9H,2-7,12H2,1H3;1H
Solution Type Neat
SMILES Cl.CC12CC3CC(C1)CC(N)(C3)C2
IUPAC Name 3-methyladamantan-1-amine;hydrochloride
Molecular Weight (g/mol) 201.74
Quantity 4 x 25 mg
Formula Weight 201.1284 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.