accessibility menu, dialog, popup

UserName

CAS RN 3304-51-6

IUPAC Name: 2-amino-5-{[(benzyloxy)carbonyl]amino}pentanoic acid
Synonyms: 2-amino-5-{[(benzyloxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 266.3
Molecular Formula: C13H18N2O4
InChi Key: VULSXQYFUHKBAN-UHFFFAOYNA-N
SMILES: NC(CCCNC(=O)OCC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results