accessibility menu, dialog, popup

UserName

H2N(R)O(R)O(R)(S)NH2(R)H2N(S)O(S)O(R)OH(S)OH(S)H2N(R)HO(S)OH(R)NH2(S)OH

CAS RN 32986-56-4

IUPAC Name: (2S,3R,4S,5S,6R)-4-amino-2-{[(1S,2S,3R,4S,6R)-4,6-diamino-3-{[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-6-(hydroxymethyl)oxane-3,5-diol
Synonyms: tobramycin obramycin tobramaxin sprc-AB01 tobramicina tobrex sulfate, tobramycin tobradistin gotabiotic tobramycin sulphate
Molecular Weight (g/mol): 467.52
Molecular Formula: C18H37N5O9
InChi Key: NLVFBUXFDBBNBW-PBSUHMDJSA-N
SMILES: NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)[C@H](N)C[C@@H]1O
Quantity 
  • (1)
  • (4)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (8)
Percent Purity 
  • (3)
  • (3)
  • (2)
Physical Form 
  • (5)
  • (2)
Type 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (5)
  • (8)
  • (3)
  • (3)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)