missing translation for 'onlineSavingsMsg'
Learn More

Tobramycin, MP Biomedicals™

Product Code. 11446225 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446225 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11446225 Supplier MP Biomedicals Supplier No. 0215692691
  • ONLINE EXCLUSIVE PRICE
    1200.00 EUR
    1139.25 EUR / 1g
    Save 60.75 EUR
    5% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Effective against gram negative bacteria, especially the Pseudomonas species. Tobramycin, MP Biomedicals is an aminoglycoside antibiotic that binds to the 70S ribosomal subunit. It inhibits translocation and hence elicits miscoding.

Tobramycin is an aminoglycoside, broad-spectrum antibiotic derived from Streptomyces tenebrarius. It is used to treat a variety of bacterial infections. Tobramycin is preferred over gentamicin for Pseudomonas aeruginosa pneumonia due to better lung penetration. It is not effective against Enterococci.

It is used in combination with other antibiotics to treat urinary tract infections, gynecologic infections, peritonitis, endocarditis, pneumonia, sepsis, respiratory infections, osteomyelitis, and other soft-tissue infections. It is a potential therapy for sinus infections.

Tobramycin inhibits bacterial protein synthesis. It works by binding to a site on bacterial 30S and 50S ribosome, preventing formation of the 70S complex. As a result, mRNA cannot be translated into protein and cell death ensues.

Specifications

Physical Form Powder
CAS 32986-56-4
Quantity 1 g
Molecular Formula C18H37N5O9
Synonym O-[3-Amino-3-deoxy-α-D-glucopyranosyl-(1?6)]-O-[2,6-diamino-2,3,6-trideoxy-α-D-ribohexopyranosyl-(1?4)]-2-deoxy-D-streptamine, Deoxykanamycin B , Gotabiotic , Nebramycin
Solubility Information Soluble in water (50mg/mL).
InChI Key NLVFBUXFDBBNBW-PBSUHMDJSA-N
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N
IUPAC Name (2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol
Molecular Weight (g/mol) 467.52
PubChem CID 36294
ChEBI CHEBI:28864
Formula Weight 467.51
Grade Not Available
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.