CAS RN 328998-53-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
inS3-54A18, MedChemExpress
MedChemExpress inS3-54A18 is a potent STAT3 inhibitor, with anti-cancer properties.