accessibility menu, dialog, popup

UserName

CAS RN 312739-90-5

H3CCCH3CH3H3CCH3H3CCCH3CH3H3CCH3Ba

CAS RN 312739-90-5

IUPAC Name: barium(2+) bis(pentamethylcyclopenta-1,3-dien-1-ide)
Synonyms: barium(2+) bis(pentamethylcyclopenta-1,3-dien-1-ide)
Molecular Weight (g/mol): 407.79
Molecular Formula: C20H30Ba
InChi Key: ZVWNPBSWYPWMIF-UHFFFAOYSA-N
SMILES: [Ba++].CC1=[C-]C(C)(C)C(C)=C1C.CC1=[C-]C(C)(C)C(C)=C1C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results