accessibility menu, dialog, popup

UserName

CH3OOBrHO

CAS RN 3111-37-3

IUPAC Name: 3-bromo-5-ethoxy-4-hydroxybenzaldehyde
Synonyms: 3-bromo-5-ethoxy-4-hydroxybenzaldehyde
Molecular Weight (g/mol): 245.07
Molecular Formula: C9H9BrO3
InChi Key: GWEFTCNMUHHQLP-UHFFFAOYSA-N
SMILES: CCOC1=CC(C=O)=CC(Br)=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results