accessibility menu, dialog, popup

UserName

CAS RN 30752-18-2

IUPAC Name: 1-bromo-4-(pentyloxy)benzene
Synonyms: 1-bromo-4-(pentyloxy)benzene
Molecular Weight (g/mol): 243.14
Molecular Formula: C11H15BrO
InChi Key: ILLQRHZDICIFRQ-UHFFFAOYSA-N
SMILES: CCCCCOC1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results