accessibility menu, dialog, popup

UserName

CAS RN 306-67-2

HHHHClClClClH2NNHHNNH2

CAS RN 306-67-2

IUPAC Name: tetrahydrogen (3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine tetrachloride
Synonyms: tetrahydrogen spermine tetrachloride
Molecular Weight (g/mol): 348.18
Molecular Formula: C10H30Cl4N4
InChi Key: XLDKUDAXZWHPFH-UHFFFAOYSA-N
SMILES: [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN
Physical Form 
  • (1)
  • (4)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)