accessibility menu, dialog, popup

UserName

NHN(R)O(S)HO(S)OOCH3NNN

CAS RN 30516-87-1

IUPAC Name: 1-[(2R,4S,5S)-4-azido-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: zidovudine aztec azt antiviral apo-zidovudine zidovudine product zidovudine preparation retrovir zidovudine (product) zidovudine EP III ZDV
Molecular Weight (g/mol): 267.25
Molecular Formula: C10H13N5O4
InChi Key: HBOMLICNUCNMMY-XLPZGREQSA-N
SMILES: CC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](CO)O2)C(=O)NC1=O
Molecular Weight (g/mol) 
  • (6)
  • (7)
Quantity 
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (6)
  • (2)
Physical Form 
  • (4)
  • (2)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (6)
  • (7)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (6)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)