missing translation for 'onlineSavingsMsg'
Learn More

3'-Azido-3'-Deoxythymidine, 97.64%, MP Biomedicals™

Product Code. 11458931 Shop All MP Biomedicals Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
missing translation for 'unitSize':
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
11458931 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11458931 missing translation for 'mfr' MP Biomedicals missing translation for 'supplierNo' 0215480791

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reverse transcriptase inhibitor active against HIV-1 virus. Soluble in water (50mg/mL)

Specifications

CAS 30516-87-1
Color White
Molecular Formula C10H13N5O4
MDL Number MFCD00006536
Quantity 1 g
Synonym azt; azidothymidine; zdv; zidovudine, 1-2r-4-azido-5-hydroxymethyl oxolan-2-yl-5-methyl-3h-pyrimidine-2,4-dione
InChI Key HBOMLICNUCNMMY-CFQLRCIDNA-N
SMILES CC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](CO)O2)C(=O)NC1=O
IUPAC Name 1-[(2R,4S,5S)-4-azido-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Weight (g/mol) 267.25
PubChem CID 455007
Formula Weight 267.2
Physical Form Powder
Chemical Name or Material 3'-Azido-3'-Deoxythymidine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.