accessibility menu, dialog, popup

UserName

CAS RN 3033-62-3

H3CNCH3ONCH3CH3

CAS RN 3033-62-3

IUPAC Name: {2-[2-(dimethylamino)ethoxy]ethyl}dimethylamine
Synonyms: {2-[2-(dimethylamino)ethoxy]ethyl}dimethylamine
Molecular Weight (g/mol): 160.26
Molecular Formula: C8H20N2O
InChi Key: GTEXIOINCJRBIO-UHFFFAOYSA-N
SMILES: CN(C)CCOCCN(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results