Learn More
Bis(2-dimethylaminoethyl) ether, 98%
Product Code. 11382327
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382327 50 g 50g
11372327 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11382327 Supplier Thermo Scientific Alfa Aesar Supplier No. L09884.18
Request Bulk or Custom Format
CAS: 3033-62-3 | C8H20N2O | 160.26 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3033-62-3
Molecular Formula C8H20N2O
Molecular Weight (g/mol) 160.26
MDL Number MFCD00059199
InChI Key GTEXIOINCJRBIO-UHFFFAOYSA-N
Synonym bis 2-dimethylaminoethyl ether, niax catalyst al, kalpur pc, toyocat ets, niax a 1, texacat zf 20, dabco bl 19i, dabco bl 11, dabco bl 19
PubChem CID 18204
IUPAC Name 2-[2-(dimethylamino)ethoxy]-N,N-dimethylethanamine
SMILES CN(C)CCOCCN(C)C
Specifications
Density 0.85
Boiling Point 179°C to 180°C
Flash Point 66°C (151°F)
Refractive Index 1.43
Quantity 50 g
UN Number UN2922
Beilstein 1739668
Formula Weight 160.26
Percent Purity 98%
Chemical Name or Material Bis(2-dimethylaminoethyl) ether