accessibility menu, dialog, popup

UserName

CAS RN 2926-27-4

FFFSOOOK

CAS RN 2926-27-4

IUPAC Name: potassium trifluoromethanesulfonate
Synonyms: potassium triflate
Molecular Weight (g/mol): 188.16
Molecular Formula: CF3KO3S
InChi Key: GLGXXYFYZWQGEL-UHFFFAOYSA-M
SMILES: [K+].[O-]S(=O)(=O)C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results