accessibility menu, dialog, popup

UserName

CAS RN 28994-41-4

OH

CAS RN 28994-41-4

IUPAC Name: 2-benzylphenol
Synonyms: phenol, 2-(phenylmethyl)-
Molecular Weight (g/mol): 184.24
Molecular Formula: C13H12O
InChi Key: CDMGNVWZXRKJNS-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)