missing translation for 'onlineSavingsMsg'
Learn More

Delegol T, TRC

Product Code. 30367838
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367838 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367838 Supplier Toronto Research Chemicals Supplier No. TRCD3899405G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxydiphenylmethane
CAS 28994-41-4
Molecular Formula C13 H12 O
Quantity 5 g
Synonym 2-Hydroxydiphenylmethane, 2-Benzylphenol, Phenol, 2-(phenylmethyl)- (9CI, ACI), 2-(Phenylmethyl)phenol (ACI), o-Cresol, α-phenyl- (6CI, 7CI, 8CI), (2-Hydroxydiphenyl)methane, 2-Benzylphenol, Delegol T, o-Benzylphenol, o-Hydroxydiphenylmethane, α-Phenyl-o-cresol
InChI Formula InChI=1S/C13H12O/c14-13-9-5-4-8-12(13)10-11-6-2-1-3-7-11/h1-9,14H,10H2
SMILES Oc1ccccc1Cc2ccccc2
IUPAC Name 2-benzylphenol
Molecular Weight (g/mol) 184.23
Formula Weight 184.0888
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.