accessibility menu, dialog, popup

UserName

NaClNONONClO

CAS RN 2893-78-9

IUPAC Name: sodium 3,5-dichloro-2,4,6-trioxo-1,3,5-triazinan-1-ide
Synonyms: sodium dichloroisocyanurate
Molecular Weight (g/mol): 219.94
Molecular Formula: C3Cl2N3NaO3
InChi Key: MSFGZHUJTJBYFA-UHFFFAOYSA-M
SMILES: [Na+].ClN1C(=O)[N-]C(=O)N(Cl)C1=O
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)