accessibility menu, dialog, popup

UserName

CH3H3CNONCH3OHNN

CAS RN 28822-58-4

IUPAC Name: 1-methyl-3-(2-methylpropyl)-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: 3 isobutyl 1 methylxanthine 3-isobutyl-1-methyxanthine IBMX
Molecular Weight (g/mol): 222.25
Molecular Formula: C10H14N4O2
InChi Key: APIXJSLKIYYUKG-UHFFFAOYSA-N
SMILES: CC(C)CN1C2=C(NC=N2)C(=O)N(C)C1=O
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (2)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (7)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)