accessibility menu, dialog, popup

UserName

CAS RN 286456-42-6

CH3CH3H3CNNNNONNNCH3FF

CAS RN 286456-42-6

IUPAC Name: 5-({[7-tert-butyl-3-(2,5-difluorophenyl)-[1,2,4]triazolo[4,3-b]pyridazin-6-yl]oxy}methyl)-1-methyl-1H-1,2,4-triazole
Synonyms: 5-({[7-tert-butyl-3-(2,5-difluorophenyl)-[1,2,4]triazolo[4,3-b]pyridazin-6-yl]oxy}methyl)-1-methyl-1,2,4-triazole
Molecular Weight (g/mol): 399.41
Molecular Formula: C19H19F2N7O
InChi Key: BQDUNOMMYOKHEP-UHFFFAOYSA-N
SMILES: CN1N=CN=C1COC1=NN2C(C=C1C(C)(C)C)=NN=C2C1=C(F)C=CC(F)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results