accessibility menu, dialog, popup

UserName

NaCH3O(R)(R)(R)(S)CH3OH3COOO(R)OH3CCH3H3COO(R)(S)CH3(S)O(R)HOHO(R)H3C(S)CH3(R)H3C

CAS RN 28643-80-3

IUPAC Name: sodium 2-[(3S,6R)-6-{[(5R,7R,9R,10R)-2-[(3'S,5'R)-5'-[(2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-2,3'-dimethyl-[2,2'-bioxolan]-5-yl]-9-methoxy-2,4,10-trimethyl-1,6-dioxaspiro[4.5]decan-7-yl]methyl}-3-methyloxan-2-yl]propanoate
Synonyms: sodium 2-[(3S,6R)-6-{[(5R,7R,9R,10R)-2-[(3'S,5'R)-5'-[(2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-2,3'-dimethyl-[2,2'-bioxolan]-5-yl]-9-methoxy-2,4,10-trimethyl-1,6-dioxaspiro[4.5]decan-7-yl]methyl}-3-methyloxan-2-yl]propanoate
Molecular Weight (g/mol): 746.96
Molecular Formula: C40H67NaO11
InChi Key: MOYOTUKECQMGHE-KKCUGXASSA-M
SMILES: [Na+].CO[C@@H]1C[C@@H](C[C@H]2CC[C@H](C)C(O2)C(C)C([O-])=O)O[C@]2(OC(C)(CC2C)C2CCC(C)(O2)C2O[C@H](C[C@@H]2C)[C@H]2O[C@@](O)(CO)[C@H](C)C[C@@H]2C)[C@@H]1C
Molecular Weight (g/mol) 
  • (1)
  • (8)
Percent Purity 
  • (1)
  • (3)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Color 
  • (5)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (3)
  • (5)
  • (1)
  • (1)
  • (8)
  • (1)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (5)
  • (5)