accessibility menu, dialog, popup

UserName

CAS RN 28230-32-2

HONNNO

CAS RN 28230-32-2

IUPAC Name: 3-hydroxy-3,4-dihydro-1,2,3-benzotriazin-4-one
Synonyms: 3-hydroxy-1,2,3-benzotriazin-4-one
Molecular Weight (g/mol): 163.14
Molecular Formula: C7H5N3O2
InChi Key: HJBLUNHMOKFZQX-UHFFFAOYSA-N
SMILES: ON1N=NC2=CC=CC=C2C1=O
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results