missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris™ HODHBt
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
HODHBt is a STAT5-SUMO protein-protein interaction inhibitor. Blocks SUMOylation of phosphorylated STAT5. Reactivates latent HIV-1 reservoir in central memory T cells (TCM). Increases viral release by ~2.7-fold at a concentration of 100 μM in combination with IL-2 compared to unstimulated cells; reduces size of latent HIV-1 reservoir in vitro.
Specifications
Specifications
| Chemical Name or Material | 3-Hydroxy-1,2,3-benzotriazin-4(3 H)-one |
| CAS | 28230-32-2 |
| Quantity | 100 mg |
| Solubility | Soluble to 100 mM in DMSO and to 50 mM in ethanol |
| Target | Antiviral Compounds |
| Molecular Formula | C7H5N3O2 |
| Formula Weight | 163.14 |
| Product Type | Inhibitor |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction