accessibility menu, dialog, popup

UserName

CAS RN 265129-71-3

H3CCH3SNONHOHO

CAS RN 265129-71-3

IUPAC Name: 2-[(4-{2-[(4-cyclohexylbutyl)(cyclohexylcarbamoyl)amino]ethyl}phenyl)sulfanyl]-2-methylpropanoic acid
Synonyms: 2-[(4-{2-[(4-cyclohexylbutyl)(cyclohexylcarbamoyl)amino]ethyl}phenyl)sulfanyl]-2-methylpropanoic acid
Molecular Weight (g/mol): 502.76
Molecular Formula: C29H46N2O3S
InChi Key: PKNYXWMTHFMHKD-UHFFFAOYSA-N
SMILES: CC(C)(SC1=CC=C(CCN(CCCCC2CCCCC2)C(=O)NC2CCCCC2)C=C1)C(O)=O
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Color 
  • (5)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results