missing translation for 'onlineSavingsMsg'
Learn More

GW7647, MedChemExpress

Product Code. 16498527
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498527 5 mg 5mg
16475888 10 mg 10mg
16472037 50 mg 50mg
16466346 100 mg 100mg
16433396 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16498527 Supplier Medchem Express Supplier No. HY13861

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GW7647 is a potent PPARα agonist, with EC50s of 6 nM, 1.1 μM, and 6.2 μM for human PPARα, PPARγ and PPARδ, respectively.

GW7647 is a potent PPARα agonist, with EC50s of 6 nM, 1.1 μM, and 6.2 μM for human PPARα, PPARγ and PPARδ, respectively.

Specifications

Chemical Name or Material GW7647
Physical Form Solid
CAS 265129-71-3
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C29H46N2O3S
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : ≥ 60 mg/mL (119.34 mM)
SMILES CC(C)(SC1=CC=C(CCN(C(NC2CCCCC2)=O)CCCCC3CCCCC3)C=C1)C(O)=O
Molecular Weight (g/mol) 502.75
Formula Weight 502.75
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.