accessibility menu, dialog, popup

UserName

CH3FNH2Cl

CAS RN 261762-85-0

IUPAC Name: 1-(2-chloro-6-fluoro-3-methylphenyl)methanamine
Synonyms: 1-(2-chloro-6-fluoro-3-methylphenyl)methanamine
Molecular Weight (g/mol): 173.62
Molecular Formula: C8H9ClFN
InChi Key: YBTHUEGENCDRSW-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C(CN)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results