accessibility menu, dialog, popup

UserName

NH4FeFeCNCNCNCNCNCN

CAS RN 25869-00-5

IUPAC Name: iron(3+) λ²-iron(2+) ammonium hexakis(iminomethanide)
Synonyms: iron(3+) λ²-iron(2+) ammonium hexakis(cyanide)
Molecular Weight (g/mol): 285.84
Molecular Formula: C6H4Fe2N7
InChi Key: ILZWGESBVHGTRX-UHFFFAOYSA-O
SMILES: [NH4+].[Fe++].[Fe+3].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results