CAS RN 258282-23-4
CAS RN 258282-23-4
IUPAC Name:
N1-(1-phenylcyclohexyl)pentane-1,5-diamine dihydrobromide
Synonyms:
N1-(1-phenylcyclohexyl)pentane-1,5-diamine dihydrobromide
Molecular Weight (g/mol):
422.25
Molecular Formula:
C17H30Br2N2
InChi Key:
ZCYUSVRXEKAQSL-UHFFFAOYSA-N
SMILES:
Br.Br.NCCCCCNC1(CCCCC1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
Tocris Bioscience™ IEM 1925 dihydrobromide
Tocris Bioscience™ IEM 1925 dihydrobromide
CAS: 258282-23-4 Molecular Formula: C17H30Br2N2