accessibility menu, dialog, popup

UserName

H3CNHOHOCH3HNO(E/Z)

CAS RN 251356-45-3

IUPAC Name: 3-{2,4-dimethyl-5-[(2-oxo-6-phenyl-2,3-dihydro-1H-indol-3-ylidene)methyl]-1H-pyrrol-3-yl}propanoic acid
Synonyms: 3-{2,4-dimethyl-5-[(2-oxo-6-phenyl-1H-indol-3-ylidene)methyl]-1H-pyrrol-3-yl}propanoic acid
Molecular Weight (g/mol): 386.45
Molecular Formula: C24H22N2O3
InChi Key: APYYTEJNOZQZNA-UHFFFAOYSA-N
SMILES: CC1=C(CCC(O)=O)C(C)=C(N1)C=C1C(=O)NC2=C1C=CC(=C2)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results