accessibility menu, dialog, popup

UserName

H3COO(R)(S)H3C(S)(S)O(S)H3C(R)F(S)OH(S)H3COCl

CAS RN 25122-46-7

IUPAC Name: (1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-1-(2-chloroacetyl)-9b-fluoro-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl propanoate
Synonyms: temovate clobetasol propionate dermovate clobex luxiq clobetasol 17-propionate olux E olux-E embeline Olux
Molecular Weight (g/mol): 466.97
Molecular Formula: C25H32ClFO5
InChi Key: CBGUOGMQLZIXBE-XGQKBEPLSA-N
SMILES: CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CCl
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results