accessibility menu, dialog, popup

UserName

OOHNHSOOFFF

CAS RN 250714-63-7

IUPAC Name: 3-phenyl-2-[3-(trifluoromethyl)benzenesulfonamido]propanoic acid
Synonyms: 3-phenyl-2-[3-(trifluoromethyl)benzenesulfonamido]propanoic acid
Molecular Weight (g/mol): 373.35
Molecular Formula: C16H14F3NO4S
InChi Key: JFDJLMGVJXHLOF-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=CC=C1)NS(=O)(=O)C1=CC=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)