accessibility menu, dialog, popup

UserName

HClCH3NH3COHO

CAS RN 2491-06-7

IUPAC Name: hydrogen 2-(dimethylamino)acetic acid chloride
Synonyms: hydrogen dimethylglycine chloride
Molecular Weight (g/mol): 139.58
Molecular Formula: C4H10ClNO2
InChi Key: FKASAVXZZLJTNX-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CC(O)=O
Molecular Weight (g/mol) 
  • (8)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results