CAS RN 2474876-09-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ iBET-BD2
Potent and selective pan-BD2 inhibitor; orally bioavailable