accessibility menu, dialog, popup

UserName

H2OClClH3COHCH3(R)(R)(S)OH(R)(S)NCH3H3CONH2OHO(S)OHOOHOHCH3(R)(R)(S)OH(R)(S)NCH3H3CONH2OHO(S)OHOOHOHHH(E)(E)

CAS RN 24390-14-5

IUPAC Name: dihydrogen bis((2E,4S,4aR,5S,5aR,6R,12aS)-2-[amino(hydroxy)methylidene]-4-(dimethylamino)-5,10,11,12a-tetrahydroxy-6-methyl-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione) ethanol hydrate dichloride
Synonyms: dihydrogen bis((2E,4S,4aR,5S,5aR,6R,12aS)-2-[amino(hydroxy)methylidene]-4-(dimethylamino)-5,10,11,12a-tetrahydroxy-6-methyl-4a,5,5a,6-tetrahydro-4H-tetracene-1,3,12-trione) ethyl alcohol hydrate dichloride
Molecular Weight (g/mol): 1025.88
Molecular Formula: C46H58Cl2N4O18
InChi Key: QDDHZFKCHWYXJF-PCLMCNLLSA-N
SMILES: [H+].[H+].O.[Cl-].[Cl-].CCO.C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H](N(C)C)C(=O)\C(=C(\N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12.C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H](N(C)C)C(=O)\C(=C(\N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12
Molecular Weight (g/mol) 
  • (6)
  • (1)
  • (5)
  • (1)
  • (5)
Quantity 
  • (3)
  • (1)
  • (2)
  • (4)
  • (3)
  • (1)
Grade 
  • (2)
  • (4)
Percent Purity 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (4)
  • (6)
  • (1)
  • (5)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (4)
  • (3)
  • (1)
  • (7)
  • (2)
  • (4)
  • (4)
  • (2)
  • (5)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)