accessibility menu, dialog, popup

UserName

CAS RN 243666-15-1

NHNNFFF

CAS RN 243666-15-1

IUPAC Name: 1-[3-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane
Synonyms: 1-[3-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane
Molecular Weight (g/mol): 245.25
Molecular Formula: C11H14F3N3
InChi Key: BMKFDQXBPCCAFY-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(N=CC=C1)N1CCCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results