missing translation for 'onlineSavingsMsg'
Learn More

1-(6-Trifluoromethyl-2-pyridyl)-1,4-diazepane, 95%, Thermo Scientific™

Product Code. 15418595
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418595 250 mg 250mg
15416547 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418595 Supplier Thermo Scientific Alfa Aesar Supplier No. H33689.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 243666-15-1
Molecular Formula C11H14F3N3
Molecular Weight (g/mol) 245.249
MDL Number MFCD12756231
InChI Key BMKFDQXBPCCAFY-UHFFFAOYSA-N
Synonym 1-3-trifluoromethyl pyridin-2-yl-1,4-diazepane, 1-3-trifluoromethyl pyrid-2-yl-1,4-diazepane, 1-3-trifluoromethyl pyridin-2-yl homopiperazine, 1-3-trifluoromethyl-2-pyridyl-1,4-diazaperhydroepine
PubChem CID 2778749
IUPAC Name 1-[3-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane
SMILES C1CNCCN(C1)C2=C(C=CC=N2)C(F)(F)F

Specifications

Refractive Index 1.521
Quantity 250 mg
UN Number UN3267
Formula Weight 245.25
Percent Purity 95%
Chemical Name or Material 1-(6-Trifluoromethyl-2-pyridyl)-1,4-diazepane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.