accessibility menu, dialog, popup

UserName

CH3NOOOOH

CAS RN 23876-15-5

IUPAC Name: 2-(2-methyl-3-nitrophenyl)acetic acid
Synonyms: (2-methyl-3-nitrophenyl)acetic acid
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: LWIOFILTAJJDLA-UHFFFAOYSA-N
SMILES: CC1=C(CC(O)=O)C=CC=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results