accessibility menu, dialog, popup

UserName

ClOOHFFF

CAS RN 2376-00-3

IUPAC Name: 2-chloro-6-(trifluoromethyl)benzoic acid
Synonyms: 2-chloro-6-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 224.56
Molecular Formula: C8H4ClF3O2
InChi Key: OHWGZCNPFKBCDJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=CC=C1Cl)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results