accessibility menu, dialog, popup

UserName

H2NFFFF

CAS RN 235106-09-9

IUPAC Name: 1-[3-fluoro-4-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[3-fluoro-4-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 193.15
Molecular Formula: C8H7F4N
InChi Key: XYWYEOHNURYGGP-UHFFFAOYSA-N
SMILES: NCC1=CC=C(C(F)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results