CAS RN 23491-44-3
CAS RN 23491-44-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress Hoechst 33258
MedChemExpress Hoechst 33258 is a fluorescent dye that emits blue fluorescence when bound to dsDNA.