Learn More
MedChemExpress Hoechst 33258
Specifications
Specifications
| Content And Storage | -20°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Quantity | 50 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 41.67 mg/mL (98.16 mM; ultrasonic and warming and heat to 60°C) |
| Molecular Weight (g/mol) | 424.5 g/mol |
| CAS | 23491-44-3 |
| Synonym | bisBenzimide H 33258 ∣ H 33258 |