accessibility menu, dialog, popup

UserName

HOOOHOO

CAS RN 2321-07-5

IUPAC Name: 3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: fluorescein uranine fluorescein stain fluorets uranin acid yellow 73 fluorescite Ful-Glo ful glo yellow fluorescein
Molecular Weight (g/mol): 332.31
Molecular Formula: C20H12O5
InChi Key: GNBHRKFJIUUOQI-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(OC3=CC(O)=CC=C3C22OC(=O)C3=CC=CC=C23)=C1
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
Grade 
  • (5)
Color 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (6)
  • (2)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)